Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ E-PAK™ Cartridges, Commercial Scale, SiliaCarb™ C-VW Activated Carbon

E-PAK™ Cartridges, Commercial Scale, SiliaCarb™ C-VW Activated Carbon is a virgin wood based catalytic activated carbon conveniently packaged in a cartridge.

$2230.00 - $3440.00

Specifications

Type Commercial Scale E-PAK Cartridge
For Use With (Application) Removal of precious metal catalysts, pigments and reaction contaminants
Color Black
Quantity 1 Pk.
pH 6 to 8
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
01-182-022
View Documents
SiliCycle™
CSCVW1650
50 x 16.5 cm
Each for $2,230.00
Add to Cart
 
01-182-021
View Documents
SiliCycle™
CSCVW16100
100 x 16.5 cm
Each for $3,440.00
Add to Cart
 
Description

Description

  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
  • Activated Carbon weight: 3.4 kg (16.5 x 50 cm), 6.5 kg (16.5 x 100 cm)
Specifications

Specifications

Commercial Scale E-PAK Cartridge
Black
6 to 8
Removal of precious metal catalysts, pigments and reaction contaminants
1 Pk.
Virgin Wood Carbon
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.