Learn More
Medchemexpress LLC Duvakitug, 10mg, Humanized IgG1-λ2 Monoclonal Antibody
Supplier: Medchemexpress LLC HYP99000610MG
Duvakitug is a humanized IgG1-λ2 monoclonal antibody targeting to TNFSF15/TL1A. Duvakitug' main expression system is CHOK1SV cells endogenously expressing glutamine synthetase (GS). Duvakitug can be used in the study of Crohn's Disease (CD).