Learn More
Medchemexpress LLC HY-125079 5mg Medchemexpress, DSP-2230 CAS:1233231-30-5 Purity:>98%
Supplier: Medchemexpress LLC HY1250795MG
Medchemexpress, HY-125079 5mg DSP-2230 CAS:1233231-30-5 DSP-2230 is a selective Nav1.7/Nav1.8 blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.