Learn More
Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Assay
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI Cotinine Drugs of Abuse Assay.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | Cotinine |
Sample Type | Urine |
Type | Reagent |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-319
|
Thermo Scientific™
0394 |
1 x 100 mL |
Each of 1 for $3,203.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. This liquid, ready-to-use assay has been field-tested on a wide range of analyzers. It is enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
Specifications
2°C to 8°C | |
Cotinine | |
Urine | |
Reagent |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.