Learn More
Grainger Disposable Bailer, HDPE, 1-1/2" Diameter, 3 ft., 24/pk
Supplier: Grainger 9MFM0
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
MonoFlex Disposable Bailer, 1000mL, Include both slow and fast flow bottom-emptying devices, Ideal for well purging and sample retrieval, 24/pk