Learn More
Siemens Healthineers Dimertest Latex Assay
The DIMERTEST™ Latex Assay is intended for the rapid qualitative or semiquantitative evaluation of circulating derivatives of cross-linked fibrin degradation products (XL-FDP) in human plasma.
Supplier: Siemens Healthineers 10445722
This item is not returnable.
View return policy
Description
A rapid agglutination assay using latex particles coated with a specific D-dimer monoclonal antibody, Dimertest is intended for the qualitative or semiquantitative evaluation of cross-linked fibrin degradation products containing D-dimers.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Hemostasis Reagent | |
D-Dimer Testing | |
60 Tests/Kit |