Learn More
Rice Lake Weighing System Scale, Hanging OS-10 10kg x 0.01kg/22lb x 0.02lb/22lb x 1/4oz Includes carrying case, 2 AA size Alkaline Batteries, manual Bottom hook fully attached, top hook removable
Supplier: Rice Lake Weighing System 130292
Medical Scale, Rice Lake Weighing Systems, Hanging OS-10 10kg x 0.01kg/22lb x 0.02lb/22lb x 1/4oz Includes carrying case, (2) AA Batteries, bottom hook fully attached, top hook removable, adjustable auto shutoff, units kg/lb/lb-oz