Learn More
BD Difco™ Glycerol
Used as a fixative, in bacterial preservation media and in the isolation and cultivation of many organisms. BD™ Difco™ Glycerol is a highly purified alcohol that helps support the growth of a wide variety of microorganisms, including mycobacterial species and fastidious bacteria.
Supplier: BD 228210
Description
Glycerol is a viscous, highly purified, fermentable alcohol often used to supplement the base of culture media (e.g. Agar). Glycerol is mixed into the solution with the culture base to support the growth of a wide variety of microorganisms.
- CAS Number: 56-81-5
- EC number: 200-289-5
- Chemical formula: C3H8O3
- Molar mass: 92.09g/mol
- Density: 1.261 g/cm3
- Solubility in water: miscible
- Viscosity: 1.412 Pa•s
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ISO/DIS, 11014 | |
15°C to 30°C | |
Highly Viscous | |
Bottle |
Colorless | |
Glycerol | |
Bacterial preservation media and in the isolation and cultivation of many organisms | |
100 g |
Safety and Handling
ShelfLife : 730 Days
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.