Learn More
Medchemexpress LLC HY-B2212 100mg Medchemexpress, Dibenzoyl Thiamine CAS:299-88-7 Purity:>98%
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HYB2212100MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medchemexpress, HY-B2212 100mg Dibenzoyl Thiamine CAS:299-88-7 Dibenzoyl Thiamine (Bentiamine), a derivative of thiamine, is rapidly absorbed into the body and converted to thiamine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.