Learn More
Electron Microscopy Sciences Precision Diamond Wafer Blade 4", Coarse grit with High Concentration, 0.012" thick
Supplier: Electron Microscopy Sciences 50265CH4
These bonded blades are constructed of an inner metal core and an outer rim, this rim is bonded with diamond particles. These blades come in high and low concentrations of diamond particles to handle various sectioning requirements. A high concentration of coarse diamond particles is for general laboratory use with low or high saw's speed.
Particles: Coarse
Concentration: High
Thickness: 0.012"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.