Learn More
Perkin Elmer US LLC Diamond Anvil Cell for the 5X Beam Condenser
Supplier:  Perkin Elmer US LLC N9302618
Encompass_Preferred
Request Bulk Order
The 5X Beam Condenser (01860794) is ideal for the analysis of microsamples. Its optical design provides optimum light-gathering power and ease of use when working with microsamples.
Catalog No. 50-211-2597
Explore available promotions