Learn More
Diamedix™ Immunosimplicity™ VZV IgG Test Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Detects IgG antibodies to Varicella-Zoster Virus antigen in human serum
Supplier: Diamedix™ 720380
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Alert:
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes: Antigen Wells, Cut-off Calibrator, Positive Control, Negative Control, Sample A Diluent, Wash S Concentrate (20X), Conjugate, Substrate HRP, Stop M Solution
Description
The Diamedix™ Immunosimplicity™ VZV IgG Test Kit is a semi-quantitative and qualitative indirect enzyme immunoassay
- Detects IgG antibodies to Varicella-Zoster Virus (VZV) antigen in human serum
- Determines a prior exposure to VZV
- Aids in the determination of acute or convalescent stage of VZV infection when evaluating paired sera
- For in vitro diagnostic use
- Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Varicella-Zoster virus IgG | |
Detects IgG Antibodies to Varicella-Zoster virus Antigen in Human Serum and aids in the determination of acute or convalescent stage of VZV infection | |
96 Tests |
Immunosimplicity™ VZV IgG Test Kit | |
Antigen wells, cut-off calibrator, positive control, negative control, sample A diluent, wash S concentrate 20X, conjugate, substrate HRP, stop M solution |