Learn More
Diamedix™ Immunosimplicity™ CMV IgG Test Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Detects IgG antibodies to cytomegalovirus in human serum
Supplier: Diamedix™ 720320
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Alert:
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes: Antigen Wells, Standards: 0 EU/mL, 10 EU/mL, 160 EU/mL, High Positive Control, Low Positive Control, Negative Control, Sample A Diluent, Wash S Concentrate (20X), Conjugate, Substrate HRP, Stop M Solution
Description
The Diamedix™ Immunosimplicity™ CMV IgG Test Kit is a qualitative and semi-quantitative indirect enzyme immunoassay
- Detects IgG antibodies to cytomegalovirus (CMV) antigen in human serum
- Aids in the assessment of the patient's immunological response to CMV and to determine the immune status of individuals, including females of child-bearing age
- The evaluation of acute and convalescent sera can aid in the diagnosis of primary infection, reactivated infection or reinfection with CMV
- For in vitro diagnostic use
- Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
CMV IgG | |
CMV IgG Test Kit | |
Antigen wells, standards: 0 EU/mL, 10 EU/mL, 160 EU/mL, high positive control, low positive control, negative control, sample A diluent, wash S concentrate 20X, conjugate, substrate HRP, stop M solution | |
96 Tests |