Learn More
Kd Medical Inc DEPC-TREATED WATER, 100ML
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Kd Medical Inc RGC3050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 100 ml bottle.