Learn More
Ampco Safety Tools Deep Well Socket, 1 in. Drive (Metric)
AMPCO™ Deep Well Socket performs in high-risk and demanding environments.
$1131.00 - $5230.00
Specifications
Product Type | Deep-well Socket Wrench |
---|---|
Drive Size | 1 in. |
Material | Aluminum Bronze |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity & Availability | |||||
19-173-676
|
Ampco Safety Tools
DW1D100MM |
100 mm |
Each for $5,230.00
|
|
|||||
19-173-706
|
Ampco Safety Tools
DW1D41MM |
41 mm |
Each for $1,131.00
|
|
|||||
19-173-707
|
Ampco Safety Tools
DW1D42MM |
42 mm |
Each for $1,131.00
|
|
|||||
19-173-708
|
Ampco Safety Tools
DW1D46MM |
46 mm |
Each for $1,240.00
|
|
|||||
19-173-709
|
Ampco Safety Tools
DW1D50MM |
50 mm |
Each for $1,412.00
|
|
|||||
19-173-710
|
Ampco Safety Tools
DW1D55MM |
55 mm |
Each for $1,691.00
|
|
|||||
19-173-711
|
Ampco Safety Tools
DW1D60MM |
60 mm |
Each for $1,722.00
|
|
|||||
19-173-712
|
Ampco Safety Tools
DW1D65MM |
65 mm |
Each for $1,960.00
|
|
|||||
19-173-713
|
Ampco Safety Tools
DW1D70MM |
70 mm |
Each for $2,360.00
|
|
|||||
19-173-714
|
Ampco Safety Tools
DW1D75MM |
75 mm |
Each for $3,000.00
|
|
|||||
19-173-715
|
Ampco Safety Tools
DW1D80MM |
80 mm |
Each for $3,400.00
|
|
|||||
19-173-716
|
Ampco Safety Tools
DW1D85MM |
85 mm |
Each for $3,660.00
|
|
|||||
19-173-717
|
Ampco Safety Tools
DW1D90MM |
90 mm |
Each for $3,810.00
|
|
|||||
19-173-718
|
Ampco Safety Tools
DW1D95MM |
95 mm |
Each for $4,280.00
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Deep-well Socket Wrench | |
1 in. | |
Aluminum Bronze | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.