Learn More
Newcomer Supply Decalcifying Solution, EDTA/Sucrose, 1 L
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Newcomer Supply 1048B
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Newcomer Supply Decalcifying Solution, EDTA/Sucrose procedure uses a chelating agent with an added TRIS buffer for gentle bone decalcification. Decalcification rate will be slower but preservation of cellular morphology is excellent and viability of staining for enzymes, immunohistochemistry antigenicity and electron microscopy is maintained. This solution is not recommended for use when proteoglycan preservation in articular cartilage is important. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.