Learn More
Glass Expansion DC Fitting Type 61 with Ratchet Connector for Analytic Jena ICP-MS
Supplier:  Glass Expansion 708032002
Encompass_Preferred
Direct Connect (DC) Nebulizer Gas Fitting Type 61 with Ratchet Connector for Analytic Jena ICP-MS. Contains Ratchet Connector Gas Line 1/4-28 x 750 Assembly (70-803-2001) and ICP Gas Inlet Fitting Type 11/32/61 (70-803-1934). For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A61- DC Nebulizers. Compatible with ICP models: Analytic Jena ICP-MS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3023