Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab CytoStat Red Fixative
SDP

An alcohol-based general purpose cytology fixative solution.

$20.35 - $118.72

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixative
For Use With (Application) Fixation
View More Specs

Alert:

Harmful if swallowed, in contact with skin, or if inhaled


Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
STLCSR1
SDP
View Documents
StatLab
CSR1
1 gal.
Each for $20.35
Case of 4 EA for $81.40
Add to Cart
 
STLCSR120ML
SDP
View Documents
StatLab
CSR120ML
120 mL Prefill
Case of 96 for $118.72
Add to Cart
 
Description

Description

  • CytoStat Red is used for diagnostic non-gynecological cytology specimens, which include, but are not limited to, sputum, washings, body fluids, scrapings, and fine needle aspiration biopsies
  • This fixative lyses red blood cells and solubilizes proteins while preserving the diagnostically relevant cells
  • Gallon, 4 per case
Specifications

Specifications

All Tissue Processors
Fixative
Fixation
Solution
Chemical Solution
Fixative
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit