Learn More
Cole-Parmer Cole-Parmer Cylindrical Closed-Dome Tank, HDPE, 30 Gal.
Supplier: Cole-Parmer 0631870
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
All tanks have a threaded screw closure with gasket that secuely fits the 6 1/4" fill well. Two mounting flats near the bottom, set 180°> apart, permit installation of a bulkhead fitting up to 2". Domed bottom drains fluid into narrow trough around base rim. High-density polyethylene tanks have a 2" bung with with cap on the top that permits access for tubing or piping. All tanks are FDA compliant.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.