Learn More
Hach Company Cylinder, Graduated, Mixing, Glass, 100 mL +-0.6 mL, 1.0 mL divisions, glass stopper #16
Supplier: Hach Company 2088642
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This glass graduated cylinder is calibrated “to contain” with a molded in base. Each includes glass penny head stopper. Graduations are in durable white enamel bake.