Learn More
Selleck Chemical LLC Cyclosporin A 50mg 59865-13-3 Cyclosporine A
Supplier: Selleck Chemical LLC S228650MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Cyclosporin A (Cyclosporine A) is an immunosuppressive agent, binds to the cyclophilin and then inhibits calcineurin with IC50 of 7 nM in a cell-free assay, widely used in organ transplantation to prevent rejection. *For Research & Development use only. Product is not intended for drug, household, or human consumption.