Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Custom TaqMan™ Gene Expression Assay, VIC
GREENER_CHOICE

Applied Biosystems Custom TaqMan Gene Expression assays, VIC, are the custom version of our predesigned TaqMan assays for quantitative real-time PCR.

Supplier:  Applied Biosystems™ 4448509

Encompass_Preferred

 View more versions of this product

Catalog No. 44-485-09

  • $685.65 / Each of 1

    Save 5%

    Reg : $722.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Description

Description

Applied Biosystems Custom TaqMan Gene Expression assays, VIC, are the custom version of our predesigned TaqMan assays for quantitative real-time PCR. Custom TaqMan Gene Expression assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5’ end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end.

Features include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
Ideal for multiplexing—combine one FAM-labeled and one VIC-labeled assay to create a duplex assay for two different gene targets in the same qPCR well
Sequences provided—with every Custom TaqMan Gene Expression you design and order

Approximate ship time
Made to order: 4–6 days in North America, 6–10 days in Europe

Pre-formulated assay
• 2 unlabeled PCR primers (900 nM each final 1X concentration)
• 1 VIC dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Tube containing a 20 x (S and M sizes) or 60 x (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store (at -15°C to -25°C.

)
Labeled PCR
Gene Expression Assay
20X
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
TaqMan™
Room Temperature
750 Reactions
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit