Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTRAX™ Cushion Trax™ Mat
GREENER_CHOICE

Cushion Trax™ features a durable vinyl diamond-plate design provides non-directional traction and is easy to clean.

Supplier:  NOTRAX™ 479R3675YB

 View more versions of this product

Catalog No. 19-086-080


Add to Cart

Description

Description

  • Long-lasting vinyl top surface made from 100% recycled materials
  • Dense closed cell PVC foam base
  • Diamond-plate surface texture provides added traction
  • Particularly suited for multi-shift operations in dry work areas
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all 4 sides
  • Certified by the NFSI (National Floor Safety Institute)
Specifications

Specifications

Black/Yellow
3 ft.
1 Ea.
Diamond plate surface texture
75 ft.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit