Learn More
TARGETMOL CHEMICALS INC CUDC-101 25MG
Supplier: TARGETMOL CHEMICALS INC T3108
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CUDC-101 is a potent inhibitor of HDAC EGFR and HER2 with IC50s of 4.4 2.4 and 15.7 nM respectively. purity: 99%