Learn More
Sartorius Cubis™ II Precision (1-place) Balance, MCE Interface
GREENER_CHOICE
Cubis™ II Precision (1-Place) Balance features fully customizable hardware, software, and connectivity
Supplier:  Sartorius MCE5202S2S00I
Request Bulk Order
 View more versions of this product
Catalog No. 14-561-215
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty and Services
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±20 mg
Precision (1-place) Balance
425 mm
95 mm
120/240 V
50/60 Hz (± 5%)
5200 g
0.1 g
±20 mg
Draft shield I - Motorized with ionizer
0.8 s
10 years
240 mm
5.4 kg
100/240 V
Product Suggestions
Product Suggestions