Learn More
CTS™ Rotea™ System + 2 year service
Supplier:  THERMO SCI LSG 3P ATTUNE A47679
Catalog No. 50-272-3573
Explore available promotions
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3,000 x g
Cell and Gene Therapy Research, Development, and Manufacturing
Counterflow Centrifugation System
50 mL to 20 L
5 to 160 mL/min.
100/240 V
44 lb. (20 kg)
Laptop
5 mL (Minimum)
16 x 11.4 x 20 in. (Body Only), 30 x 11.4 x 20 in. (with Bag Poles)
Product Suggestions
Product Suggestions