Learn More
Mitegen Crystal Mount Alteration
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Mitegen HAD1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Changed length of crystal mounts to decrease interference with high chi-angles, modification to 1 box of 20 mounts/loops/meshes(mounts ordered separately)