Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kodiak™ Crusade CT WP 6 in. Black Boot
Offers a full-grain waterproof leather upper, waterproof membrane, an ASTM approved composite toe, slip and oil-resistant rubber outsole, and electrical hazard protection to help protect your feet, and confidently tackle tough days or terrain on the job-site.
Supplier: Kodiak™ K4NKBK 110 W
Description
- Full-grain waterproof leather upper with waterproof membrane construction
- KODIAK™ SOS Smell Out Science™ odor-control technology
- Abrasion-resistant leather toe guard offers added durability
- Full leather bellows tongue
- Removable, comfort trainer perforated EVA insole
- Kodiak™ Ultimate SR and oil-resistant rubber outsole with EVA midsole offers maximum comfort
Specifications
Black | |
11W | |
Yes | |
Boot |
Leather | |
Men's | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction