Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crocetin, free acid, >90%, MP Biomedicals™

Catalog No. ICN19354325
Click to view available options
Quantity:
1 mg
5 mg
25 mg

Chemical Identifiers

CAS 27876-94-4
Molecular Formula C20H24O4
Molecular Weight (g/mol) 328.408
InChI Key PANKHBYNKQNAHN-MQQNZMFNSA-N
Synonym crocetin, trans-crocetin, 8,8'-diapocarotenedioic acid, natural yellow 6, 8,8'-diapocarotene-8,8'-dioic acid, 8,8'-diapo-psi,psi-carotenedioic acid, ccris 7484, unii-20tc155l9c, 2e,4e,6e,8e,10e,12e,14e-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid, .alpha.-crocetin
PubChem CID 5281232
ChEBI CHEBI:3918
IUPAC Name (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
SMILES CC(=CC=CC=C(C)C=CC=C(C)C(=O)O)C=CC=C(C)C(=O)O

Specifications

Color Orange
Assay Percent Range >90%
Quantity 25 mg
Formula Weight 328.4
Physical Form Powder
Chemical Name or Material Crocetin, free acid
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.