Learn More
ACROBiosystems Coxsackievirus A16 VP0 500ug
Supplier: ACROBiosystems VP0C52H3500UG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Coxsackievirus A16 (strain G-10) VP0 Protein His Tag (VP0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 2 - Gln 323 (Accession # Q65900-1).