Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ glutaGRO Supplement
Stabilized dipeptide form of L-Glutamine
Supplier: Corning™ 25015CI
This item is not returnable.
View return policy
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- 200 mM solution (100x), with 8.5g/L NaCl
- Does not degrade in liquid media during storage or incubation
- Provides superior results in several applications
- Quantity: 1 x 100mL
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
2° to 30°C | |
glutagro Supplement | |
Plastic Bottle | |
1 x 100 mL |
Liquid | |
Sterile-filtered | |
5 to 6 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction