Learn More
Haztech Systems, Inc Hazcat Copper Wire Test Tool (Chlorine Hot Wire)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Haztech Systems, Inc HW4017
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Hazcat's Copper Wire Test Tool is used to test for halogenated hydrocarbons. Prior to each use the wire is cleaned by heating and immersing in methanol. Instructions are provided with the tool. Methanol (RE2325) and test tubes can be purchased separately.