Learn More
Copper Nickel wire, 0.075mm (0.003in) dia, Thermo Scientific Chemicals
Resistance alloy, negative lead for type J thermocouple
$37.25 - $77.00
Specifications
Chemical Name or Material | Copper Nickel wire |
---|---|
Name Note | 0.075mm (0.003 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Cu:Ni 55:45 wt% |
MDL Number | MFCD00801098 |
Synonym | Constantan |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA43269H4
|
Thermo Scientific Chemicals
043269H4 |
100 m |
Each for $37.25
Order in Bulk
Request bulk or custom formats |
|
|||||
AA43269H9
|
Thermo Scientific Chemicals
043269H9 |
500 m |
Each for $77.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Resistance alloy, negative lead for type J thermocouple
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper Nickel wire | |
Cu:Ni 55:45 wt% | |
Constantan | |
Wire |
0.075mm (0.003 in.) dia. | |
MFCD00801098 | |
≈0.04g/m |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.