Learn More
World Precision Instrument Constant Current Stimulus Isolator-A365RC
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument A365RC
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Activated by conventional logic-level commands, Model A365 can be gated by any pulse generator, stimulator or computer output with automated bipolar pulsing for zero net charge on biological preparations.
To learn more about our warranty options, click here.