Learn More
Waters Corp Conductivity Detector OQ/PQ Solution
Supplier: Waters Corp 700002309
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Conductivity Detector OQ/PQ Solution is intended to be used to verify that the 432 Conductivity Detector-based HPLC systems meet operation standards. This kit is designed for use by a Waters certified engineer or user as part of a performance maintenance or qualification activity. Protocols in the use of this kit are available via the purchase of Waters qualification services or self directed qualification training and license.
The kit contains:
Qty 2 Flow Cell Constant Solution: 1 mM potasium chloride salt in water, 10 mL in an ampule.
Qty 1 OQ/PQ Solution, 500 ppm chloride, 1000 ppm bromide in water, 10mL in an ampule.
Store at room temperature.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.