Learn More
Chemglass Life Sciences Condenser, West, 200mm, 19/22 Joint, 110mm Jacket Length
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG122023
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Condenser, West, 200mm, 19/22 Joint, 110mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.