Learn More
Ace Glass, Inc. Condenser, Liebig, 250mm, 19/22
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 926115
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
250mm Liebig condenser with straight inner tube sealed through the jacket, and 19/22 joint. This condenser is acceptable for use with Abderhalden or Mini-Lab moisture test apparatus.