Learn More
Scientific Glass & Instrument Inc CONDENSER IOWA STATE GRAHAM
Supplier: Scientific Glass & Instrument Inc 6072B4
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Condenser Graham. 200mm, 28/15 Joint. Suitable For Use With Iowa State Citric Oil Distillation Apparatus (alt # 21200-2