Learn More
Ace Glass, Inc. Condenser, Graham, 300mm, 24/40
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 597914
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
300mm Graham condenser with standard taper 24/40 outer joint at top. Hose connections are for 3/8 I.D. tubing.