Learn More
Ace Glass, Inc. Condenser, Graham, 200mm, 24/40
Supplier: Ace Glass, Inc. 597912
200mm Graham condenser with standard taper 24/40 outer joint at top. Hose connections are for 3/8 I.D. tubing.