Learn More
Chemglass Life Sciences Condenser, Dewar Type, 24/40 Joint, 350 x 75mm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1209A28
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Condenser, Dewar Type, 24/40 Joint, 350 x 75mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has an outer standard taper joint set at a 75º angle to the condenser body and a lower inner drip tip joint, both being the same size.