Learn More
Chemglass Life Sciences Condenser, Liebig, 200mm, 19/22 Top Outer, 19/22 Lower Inner, 110mm Jacket Length
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1218A22
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Condenser, Liebig, 200mm, 19/22 Top Outer, 19/22 Lower Inner, 110mm Jacket Length. Small scale Liebig condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.