Learn More
Ace Glass, Inc. Condenser, Allihn, 300mm, 24/40 st joints top & bottom
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 594514
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
300mm Allihn condenser with approximately one bulb per 50mm. Features 24/40 standard taper joints at top and bottom.