Learn More
TARGETMOL CHEMICALS INC COMPLEMENT C5-IN-1 5MG
Supplier: TARGETMOL CHEMICALS INC T10863
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Complement C5-IN-1 is a small-molecule inhibitor of complement component 5 protein (C5). purity: 99%