Learn More
Dentec Safety™ Comfort-Air™ 400NxMD Half-Mask Elastomeric Respirator with N95 Filters
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Comfort-Air™NxMD white half-mask respirators provide protection against non-oil based particulate aerosols and COVID viruses.
Supplier: Dentec Safety™ 4MLNXMDN95
Description
- Facepiece low-profile design fits under industrial style and medical style face shields
- Extended field of vision
- No exhalation valve ensures the user's exhaled breath does not exit the mask
- Rolled sealing flange provides comfort and improves fitting capabilities
- Does not require user to adjust straps excessively tight
- Textured sealing area reduces risk of the mask slipping on the face
- White color allows for written personalization and also makes dirt or other markings more visible to simplify the cleaning and disinfecting process
- Designed to allow for the proper fitting of most safety & prescription eyewear and goggles
- Ultra light harness assembly provides additional comfort
- Cradle suspension fits comfortably under suspension systems for hard hats, grinding & welding helmets
- 8- position adjustment
Specifications
White | |
Thermoplastic Elastomer | |
Medium/Large | |
Thermoplastic Elastomer | |
4-ML-NXMD-N95 |
N95 | |
Half-Mask Respirator | |
No | |
Yes | |
8-point |