Learn More
Dentec Safety™ Comfort-Air™ 400Nx Half-Mask Elastomeric Respirator with P100 Filters
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Black half-mask respirators with P100 filter cartridges do not have exhalation valves and are NIOSH approved. Provides protection against non-oil based particulate aerosols and COVID viruses.
Supplier: Dentec Safety™ 4SMNXLP0
Description
- Facepiece low profile design fits under industrial style and medical style face shields
- Also provides extended field of vision
- Rolled sealing flange provides superior comfort and improves fitting capabilities
- Does not require user to adjust straps excessively tight
- Textured sealing area reduces risk of the mask slipping on the face
- Mask is designed to allow for the proper fitting of most safety & prescription eyewear and goggles
- Ultra light harness assembly provides additional comfort
- Cradle suspension fits comfortably under suspension systems for hard hats, grinding & welding helmets
- 8-position adjustment allows you to adjust cartridge to three different positions
Specifications
Black | |
Thermoplastic Elastomer | |
Small/Medium | |
Thermoplastic Elastomer | |
4-SM-NX-LP0 |
P100 | |
Half-Mask Respirator | |
No | |
Yes | |
8-point |