Learn More
Dentec Safety™ Comfort-Air™Nx Filter Pad assembly For N95 Filter Pads
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Dentec Comfort-Air™Nx filter pad assembly for N95 filter pads. Suitable For COVID virus in accordance with CDC guidelines and non-oil based aerosols.
Supplier: Dentec Safety™ 14668BSET
Description
- Ultra thin design will fit under any grinding shield or welding helmet
- Will not obstruct field of vision
- Holder has spacers that create a space between the filter material and the holder
- This allows the user to access the entire filter material increasing filter life while reducing breathing resistance and increasing productivity
- Allows the user to replace the filter when required
- Economical solution and alternative to disposable respirators
- 'Saf-Twist' design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator facepiece
Specifications
Reusable | |
No | |
4 Pk. |