Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 10, Ricca Chemical
SDP

Supplier:  Ricca Chemical Company 2230.58

 View more versions of this product

Catalog No. R2230500250


Add to Cart

Description

Description

Specifications

Specifications

Color Standard, APHA/Hazen (Platinum-Cobalt), Color=10
250 mL
99.91, 0.09, 0.0026, 0.002
16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
<1
100°C
Liquid
Amber Glass Bottle
99.91, 0.09, 0.0026, 0.002
Brown/Yellow
1.001
0°C
Miscible with water
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit