Learn More
Creative Biomart Recombinant CMV Pp52 Antigen, GST-tagged
Supplier: Creative Biomart CMV034
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The E. coli derived 51 kDa recombinant protein contains the CMV Pp52 (UL44) immunodominant regions, 202-434 amino acids. Recombinant CMV-Pp52 is fused to a 26 kDa GST tag.