Learn More
Newcomer Supply Cytomegalovirus (CMV) Control Slides, 10/set
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Newcomer Supply 3240A
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Newcomer Supply Cytomegalovirus (CMV) Control Slides are for the positive immunohistochemical staining of cytomegalovirus, a member of the herpes virus group, including herpes simplex virus types 1 and 2, varicella zoster virus, and Epstein Barr virus. Cytomegalovirus (CMV) Control Slides contain sections of positive staining lung and negative staining myometrium.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.